Learn More
Medchemexpress LLC Propargyl-PEG1-NHS ester | 1174157-65-3 | 99.7% | 225.20 g/mol | C10H11NO5 | 250 MG
SDP
Catalog No. 5000203745
Encompass_Preferred
A non-cleavable one-unit polyethylene glycol (PEG) linker supplied as an NHS ester with a terminal propargyl (alkyne) group. Designed for click chemistry (CuAAC) and antibody-agent conjugation, it is provided as a colorless to light-yellow solid-liquid mixture with high purity and recommended cold storage.
  • One-unit PEG linker for minimal spacer length.
  • Terminal alkyne enables copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • NHS ester allows efficient coupling to primary amines.
  • High purity suitable for conjugation chemistry.
  • Cold storage recommended for long-term stability.
Catalog No. 50-002-03745 Supplier Medchemexpress LLC Supplier No. HY126513250MG
A non-cleavable one-unit polyethylene glycol (PEG) linker supplied as an NHS ester with a terminal propargyl (alkyne) group. Designed for click chemistry (CuAAC) and antibody-agent conjugation, it is provided as a colorless to light-yellow solid-liquid mixture with high purity and recommended cold storage.
  • One-unit PEG linker for minimal spacer length.
  • Terminal alkyne enables copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • NHS ester allows efficient coupling to primary amines.
  • High purity suitable for conjugation chemistry.
  • Cold storage recommended for long-term stability.