Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-PEG1-NHS ester | 1174157-65-3 | 99.7% | 225.20 g/mol | C10H11NO5 | 5 G
SDP

Catalog No. 5000202456
Encompass_Preferred

Propargyl-PEG1-NHS ester is a short PEG linker bearing a terminal propargyl (alkyne) group and an N-hydroxysuccinimide (NHS) ester for selective amine conjugation. It is used in click chemistry and bioconjugation workflows, including antibody-drug conjugate synthesis.

  • Provides a short, flexible PEG spacer to reduce steric hindrance
  • Contains an NHS ester for efficient amine coupling
  • Has a terminal alkyne for copper-catalyzed azide-alkyne cycloaddition
  • High purity suitable for research applications
  • Soluble in DMSO and stable when stored at low temperatures

Catalog No. 50-002-02456 Supplier Medchemexpress LLC Supplier No. HY1265135G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-PEG1-NHS ester is a short PEG linker bearing a terminal propargyl (alkyne) group and an N-hydroxysuccinimide (NHS) ester for selective amine conjugation. It is used in click chemistry and bioconjugation workflows, including antibody-drug conjugate synthesis.

  • Provides a short, flexible PEG spacer to reduce steric hindrance
  • Contains an NHS ester for efficient amine coupling
  • Has a terminal alkyne for copper-catalyzed azide-alkyne cycloaddition
  • High purity suitable for research applications
  • Soluble in DMSO and stable when stored at low temperatures

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.