Learn More
Medchemexpress LLC Propargyl-PEG1-SS-PEG1-C2-Boc | 1807518-78-0 | >96.0% | 320.46 g/mol | C14H24O4S2 | 5 MG
SDP
Catalog No. 5000202111
Encompass_Preferred
Propargyl-PEG1-SS-PEG1-C2-Boc is a cleavable PEG-based linker that contains a propargyl alkyne group, a disulfide (SS) bond, and a Boc-protected terminus. It is used in bioconjugation workflows such as antibody-drug conjugate (ADC) and PROTAC linker synthesis, and enables click chemistry (CuAAC) and redox-sensitive payload release.
  • Cleavable disulfide bond enabling redox-sensitive release.
  • Terminal propargyl group for copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • Boc-protected terminus allows orthogonal deprotection and functionalization.
  • One-unit PEG spacer improves solubility and linker flexibility.
  • Supplied in small research-scale quantities for conjugation workflows.
Catalog No. 50-002-02111 Supplier Medchemexpress LLC Supplier No. HY1306905MG
Propargyl-PEG1-SS-PEG1-C2-Boc is a cleavable PEG-based linker that contains a propargyl alkyne group, a disulfide (SS) bond, and a Boc-protected terminus. It is used in bioconjugation workflows such as antibody-drug conjugate (ADC) and PROTAC linker synthesis, and enables click chemistry (CuAAC) and redox-sensitive payload release.
  • Cleavable disulfide bond enabling redox-sensitive release.
  • Terminal propargyl group for copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • Boc-protected terminus allows orthogonal deprotection and functionalization.
  • One-unit PEG spacer improves solubility and linker flexibility.
  • Supplied in small research-scale quantities for conjugation workflows.