Learn More
Medchemexpress LLC Propargyl-PEG10-acid | 2055022-18-7 | 98.0% | 524.60 | 100 MG
SDP
Catalog No. 5000349479
Encompass_Preferred
Propargyl-PEG10-acid is a PEG derivative featuring a propargyl group and a terminal carboxylic acid. It serves as a click chemistry reagent, utilizing an Alkyne group to engage in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Azide groups. For research use only.
  • PEG derivative with propargyl and terminal carboxylic acid groups
  • Functions as a click chemistry reagent
  • Contains an Alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Reacts with molecules possessing Azide groups
Catalog No. 50-003-49479 Supplier Medchemexpress LLC Supplier No. HY140020100MG
Propargyl-PEG10-acid is a PEG derivative featuring a propargyl group and a terminal carboxylic acid. It serves as a click chemistry reagent, utilizing an Alkyne group to engage in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Azide groups. For research use only.
  • PEG derivative with propargyl and terminal carboxylic acid groups
  • Functions as a click chemistry reagent
  • Contains an Alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Reacts with molecules possessing Azide groups