Learn More
Medchemexpress LLC Propargyl-PEG10-acid | 2055022-18-7 | 98.0% | 524.60 | 250 MG
SDP
Catalog No. 5000348789
Encompass_Preferred
Propargyl-PEG10-acid is a PEG derivative containing a propargyl group with a terminal carboxylic acid. It functions as a click chemistry reagent, utilizing an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing azide groups.
  • PEG derivative
  • Contains a propargyl group
  • Features a terminal carboxylic acid
  • Functions as a click chemistry reagent
  • Enables copper-catalyzed azide-alkyne cycloaddition with azide groups
Catalog No. 50-003-48789 Supplier Medchemexpress LLC Supplier No. 250MGHY140020
Propargyl-PEG10-acid is a PEG derivative containing a propargyl group with a terminal carboxylic acid. It functions as a click chemistry reagent, utilizing an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing azide groups.
  • PEG derivative
  • Contains a propargyl group
  • Features a terminal carboxylic acid
  • Functions as a click chemistry reagent
  • Enables copper-catalyzed azide-alkyne cycloaddition with azide groups