Learn More
Medchemexpress LLC Propargyl-PEG10-acid | 2055022-18-7 | 98.0% | 524.60 | 50 MG
SDP
Catalog No. 5000349766
Encompass_Preferred
Propargyl-PEG10-acid is a PEG derivative that contains a propargyl group with a terminal carboxylic acid. It functions as a click chemistry reagent, featuring an alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing azide groups.
  • PEG derivative
  • Contains a propargyl group with a terminal carboxylic acid
  • Functions as a click chemistry reagent
  • Features an alkyne group
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
Catalog No. 50-003-49766 Supplier Medchemexpress LLC Supplier No. HY14002050MG
Propargyl-PEG10-acid is a PEG derivative that contains a propargyl group with a terminal carboxylic acid. It functions as a click chemistry reagent, featuring an alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing azide groups.
  • PEG derivative
  • Contains a propargyl group with a terminal carboxylic acid
  • Functions as a click chemistry reagent
  • Features an alkyne group
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions