Learn More
Medchemexpress LLC Propargyl-PEG10-acid | 2055022-18-7 | 98.0% | 524.60 | 500 MG
SDP
Catalog No. 5000349852
Encompass_Preferred
Propargyl-PEG10-acid is a PEG derivative that features a propargyl group along with a terminal carboxylic acid. It functions as a click chemistry reagent, containing an Alkyne group, and is capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules that possess Azide groups.
  • PEG derivative
  • Features a propargyl group
  • Contains a terminal carboxylic acid
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Suitable for PROTAC synthesis
  • Used as a biochemical assay reagent
Catalog No. 50-003-49852 Supplier Medchemexpress LLC Supplier No. HY140020500MG
Propargyl-PEG10-acid is a PEG derivative that features a propargyl group along with a terminal carboxylic acid. It functions as a click chemistry reagent, containing an Alkyne group, and is capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules that possess Azide groups.
  • PEG derivative
  • Features a propargyl group
  • Contains a terminal carboxylic acid
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Suitable for PROTAC synthesis
  • Used as a biochemical assay reagent