Learn More
Medchemexpress LLC HY-118764 100mg Medchemexpress, Propargyl-PEG2-acid CAS:1859379-85-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY118764100MG
Encompass_Preferred
Medchemexpress, HY-118764 100mg Propargyl-PEG2-acid CAS:1859379-85-3 Propargyl-PEG2-acid is a non-cleavable 2 unit PEG ADC linker used in the synthesis of antibody-drug conjugates (ADCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0921