Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-PEG2-amine | 944561-44-8 | 98.9% | 143.18 g/mol | C7H13NO2 | 500 MG
SDP

Catalog No. 5000204114
Encompass_Preferred

Propargyl-PEG2-amine is a PEG-based linker used as a non-cleavable linker in antibody-drug conjugate (ADC) synthesis and as a PEG-based PROTAC linker. It is supplied as a liquid reagent for bioconjugation and medicinal chemistry applications.

  • Non-cleavable ADC and PROTAC linker for bioconjugation.
  • PEG2 spacer with propargyl and amine functional groups.
  • High purity (98.9%) suitable for synthesis.
  • Liquid appearance, colorless to light yellow.
  • Molecular weight 143.18 g/mol; formula C7H13NO2.
  • Storage: refrigerate at 4°C, protect from light; in solvent store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-04114 Supplier Medchemexpress LLC Supplier No. HYW051634500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-PEG2-amine is a PEG-based linker used as a non-cleavable linker in antibody-drug conjugate (ADC) synthesis and as a PEG-based PROTAC linker. It is supplied as a liquid reagent for bioconjugation and medicinal chemistry applications.

  • Non-cleavable ADC and PROTAC linker for bioconjugation.
  • PEG2 spacer with propargyl and amine functional groups.
  • High purity (98.9%) suitable for synthesis.
  • Liquid appearance, colorless to light yellow.
  • Molecular weight 143.18 g/mol; formula C7H13NO2.
  • Storage: refrigerate at 4°C, protect from light; in solvent store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.