Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-PEG2-amine | 944561-44-8 | 99.6% | 143.18 g/mol | C7H13NO2 | 5 G
SDP

Catalog No. 5000203082
Encompass_Preferred

Propargyl-PEG2-amine is a short polyethylene glycol (PEG2) linker with a terminal propargyl (alkyne) group and a primary amine, used to introduce alkyne handles for click chemistry and bioconjugation.

  • Short PEG2 spacer with terminal alkyne and primary amine.
  • Suitable for click chemistry (CuAAC) and other bioconjugation reactions.
  • Molecular weight 143.18 g/mol; formula C7H13NO2.
  • Offered as a 5 g powder; store at 4°C, protect from light, under nitrogen.
  • CAS number 944561-44-8 for unambiguous identification.

Catalog No. 50-002-03082 Supplier Medchemexpress LLC Supplier No. HYW0516345G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-PEG2-amine is a short polyethylene glycol (PEG2) linker with a terminal propargyl (alkyne) group and a primary amine, used to introduce alkyne handles for click chemistry and bioconjugation.

  • Short PEG2 spacer with terminal alkyne and primary amine.
  • Suitable for click chemistry (CuAAC) and other bioconjugation reactions.
  • Molecular weight 143.18 g/mol; formula C7H13NO2.
  • Offered as a 5 g powder; store at 4°C, protect from light, under nitrogen.
  • CAS number 944561-44-8 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.