Learn More
Medchemexpress LLC Propargyl-peg2-beta-d-glucose | 2353409-73-9 | 98.0% | C13H22O8 | 10MG
SDP
Catalog No. 5000261298
Encompass_Preferred
Propargyl-PEG2-beta-D-glucose is an alkyne-functionalized PEG2 linker designed for PROTAC construction and bioconjugation. It offers a terminal propargyl group for click chemistry and a beta-D-glucose-derived spacer to improve solubility and biological compatibility, enabling efficient conjugation in research-scale synthetic workflows.
  • Provides terminal alkyne for copper-catalyzed click reactions.
  • Contains a two-unit polyethylene glycol spacer for solubility and flexibility.
  • Features a beta-D-glucose-derived moiety for biological compatibility.
  • Supplied at high purity suitable for synthetic applications.
  • Available in small pack sizes for research-scale synthesis.
Catalog No. 50-002-61298 Supplier Medchemexpress LLC Supplier No. HY14113110MG
Propargyl-PEG2-beta-D-glucose is an alkyne-functionalized PEG2 linker designed for PROTAC construction and bioconjugation. It offers a terminal propargyl group for click chemistry and a beta-D-glucose-derived spacer to improve solubility and biological compatibility, enabling efficient conjugation in research-scale synthetic workflows.
  • Provides terminal alkyne for copper-catalyzed click reactions.
  • Contains a two-unit polyethylene glycol spacer for solubility and flexibility.
  • Features a beta-D-glucose-derived moiety for biological compatibility.
  • Supplied at high purity suitable for synthetic applications.
  • Available in small pack sizes for research-scale synthesis.