Learn More
Medchemexpress LLC Propargyl-PEG2-bromi 250mg
SDP
Catalog No. 5000204574
Encompass_Preferred
Propargyl-PEG2-bromide is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1] Propargyl-PEG2-bromide is a click chemistry reagent it contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups
Catalog No. 50-002-04574 Supplier Medchemexpress LLC Supplier No. 250MGHYW096164
Propargyl-PEG2-bromide is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1] Propargyl-PEG2-bromide is a click chemistry reagent it contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups