Learn More
Medchemexpress LLC Propargyl-PEG2-CH2COOH | 944561-46-0 | 95.0% | 202.20 | 100 MG
SDP
Catalog No. 5000404083
Encompass_Preferred
Propargyl-PEG2-CH2COOH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Peg-based PROTAC linker
  • Click chemistry reagent
  • Contains an Alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups
Catalog No. 50-004-04083 Supplier Medchemexpress LLC Supplier No. HY41922100MG
Propargyl-PEG2-CH2COOH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Peg-based PROTAC linker
  • Click chemistry reagent
  • Contains an Alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups