Learn More
Medchemexpress LLC Propargyl-PEG2-CH2COOH | 944561-46-0 | ≥95.0% | 202.20 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY41922
Encompass_Preferred
Propargyl-PEG2-CH2COOH is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group that enables it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups.
  • Used in the synthesis of PROTACs.
  • Contains an Alkyne group.
  • Can undergo copper-catalyzed azide-alkyne cycloaddition with Azide groups.
Catalog No. 50-004-02356