Learn More
Medchemexpress LLC Propargyl-PEG2-MS | 943726-01-0 | 95.0% | 250 MG
SDP
Catalog No. 5000341460
Encompass_Preferred
Propargyl-PEG2-MS is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Participates in copper-catalyzed azide-alkyne cycloaddition with molecules possessing azide groups
Catalog No. 50-003-41460 Supplier Medchemexpress LLC Supplier No. 250MGHY130584
Propargyl-PEG2-MS is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Participates in copper-catalyzed azide-alkyne cycloaddition with molecules possessing azide groups