Learn More
Medchemexpress LLC Propargyl-PEG2-NHS ester | 2512228-06-5 | ≥98.0% | 269.25 g/mol | 100 MG
SDP
Catalog No. 5000351975
Encompass_Preferred
This PEG-based linker is designed for the synthesis of proteolysis-targeting chimeras (PROTACs), which are molecules that utilize the ubiquitin-proteasome system to degrade target proteins. It functions as a click chemistry reagent, featuring an alkyne group that enables copper-catalyzed azide-alkyne cycloaddition with azide-containing molecules.
  • Functions as a linker in targeted protein degradation research.
  • Acts as a click chemistry reagent.
  • Contains an alkyne functional group for specific reactions.
  • Can undergo copper-catalyzed azide-alkyne cycloaddition.
  • Enables conjugation with azide-containing molecules.
  • Offers high purity for reliable experimental results.
Catalog No. 50-003-51975 Supplier Medchemexpress LLC Supplier No. HY138734100MG
This PEG-based linker is designed for the synthesis of proteolysis-targeting chimeras (PROTACs), which are molecules that utilize the ubiquitin-proteasome system to degrade target proteins. It functions as a click chemistry reagent, featuring an alkyne group that enables copper-catalyzed azide-alkyne cycloaddition with azide-containing molecules.
  • Functions as a linker in targeted protein degradation research.
  • Acts as a click chemistry reagent.
  • Contains an alkyne functional group for specific reactions.
  • Can undergo copper-catalyzed azide-alkyne cycloaddition.
  • Enables conjugation with azide-containing molecules.
  • Offers high purity for reliable experimental results.