Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-peg2-oh | 7218-43-1 | 99.66% | 5 G
SDP

Catalog No. 5000340332
Encompass_Preferred

Propargyl-PEG2-OH is a PEG-based PROTAC linker that can be used in the synthesis of Thalidomide-O-PEG2-propargyl. This compound is a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups. It has a molecular weight of 144.17 and a formula of C7H12O3. It appears as a colorless to light yellow liquid with a density of 1.06 g/cm3 and a purity of 99.66%.

  • Peg-based protac linker
  • Utilized in the synthesis of thalidomide-o-peg2-propargyl
  • Click chemistry reagent
  • Contains an alkyne group for cuaac reactions
  • Intended for research use only
  • Soluble in ethanol (≥ 100 mg/ml)

Catalog No. 50-003-40332 Supplier Medchemexpress LLC Supplier No. HY1305415G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-PEG2-OH is a PEG-based PROTAC linker that can be used in the synthesis of Thalidomide-O-PEG2-propargyl. This compound is a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups. It has a molecular weight of 144.17 and a formula of C7H12O3. It appears as a colorless to light yellow liquid with a density of 1.06 g/cm3 and a purity of 99.66%.

  • Peg-based protac linker
  • Utilized in the synthesis of thalidomide-o-peg2-propargyl
  • Click chemistry reagent
  • Contains an alkyne group for cuaac reactions
  • Intended for research use only
  • Soluble in ethanol (≥ 100 mg/ml)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.