Learn More
Medchemexpress LLC Propargyl-PEG2-OH | 7218-43-1 | 99.7% | 25 G
SDP
Catalog No. 5000342809
Encompass_Preferred
Propargyl-PEG2-OH is a versatile PEG-based PROTAC linker. This compound is instrumental in the synthesis of Thalidomide-O-PEG2-propargyl and serves as a key reagent in click chemistry. Its unique Alkyne group facilitates efficient copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups, enabling broad applications in chemical synthesis and biochemical research.
  • Functions as a PEG-based PROTAC linker
  • Utilized in the synthesis of Thalidomide-O-PEG2-propargyl
  • Acts as a click chemistry reagent
  • Contains an Alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules
Catalog No. 50-003-42809 Supplier Medchemexpress LLC Supplier No. 25GHY130541
Propargyl-PEG2-OH is a versatile PEG-based PROTAC linker. This compound is instrumental in the synthesis of Thalidomide-O-PEG2-propargyl and serves as a key reagent in click chemistry. Its unique Alkyne group facilitates efficient copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups, enabling broad applications in chemical synthesis and biochemical research.
  • Functions as a PEG-based PROTAC linker
  • Utilized in the synthesis of Thalidomide-O-PEG2-propargyl
  • Acts as a click chemistry reagent
  • Contains an Alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules