Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-PEG24-amine | 00-00-0 | 98.0% | 1112.34 | C51H101NO24 | 5 MG
SDP

Catalog No. 5000223301
Encompass_Preferred

Propargyl-PEG24-amine is a PEG24-based linker bearing a terminal propargyl (alkyne) group for copper-catalyzed azide-alkyne cycloaddition (CuAAC). It is intended for PROTAC synthesis, bioconjugation, and other applications requiring a hydrophilic PEG spacer with a reactive alkyne handle. Supplied as a white to off-white solid of high purity; store under nitrogen at -20 °C for long-term storage (in solution: -80 °C up to 6 months, -20 °C up to 1 month).

  • PEG24 spacer provides flexible hydrophilic linker
  • Terminal propargyl group enables CuAAC click chemistry
  • High purity (98.0%) for synthesis and conjugation
  • Molecular weight ~1112.34 Da suitable for spacing applications
  • Solid form simplifies storage and handling
  • Recommended storage under nitrogen to maintain stability

Catalog No. 50-002-23301 Supplier Medchemexpress LLC Supplier No. HY1400355MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-PEG24-amine is a PEG24-based linker bearing a terminal propargyl (alkyne) group for copper-catalyzed azide-alkyne cycloaddition (CuAAC). It is intended for PROTAC synthesis, bioconjugation, and other applications requiring a hydrophilic PEG spacer with a reactive alkyne handle. Supplied as a white to off-white solid of high purity; store under nitrogen at -20 °C for long-term storage (in solution: -80 °C up to 6 months, -20 °C up to 1 month).

  • PEG24 spacer provides flexible hydrophilic linker
  • Terminal propargyl group enables CuAAC click chemistry
  • High purity (98.0%) for synthesis and conjugation
  • Molecular weight ~1112.34 Da suitable for spacing applications
  • Solid form simplifies storage and handling
  • Recommended storage under nitrogen to maintain stability

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.