Learn More
Medchemexpress LLC Propargyl-PEG24-amine | 98.0% | 1112.34 | 25 MG
SDP
Catalog No. 5000350148
Encompass_Preferred
Propargyl-PEG24-amine is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Click chemistry reagent
  • Contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
Catalog No. 50-003-50148 Supplier Medchemexpress LLC Supplier No. 25MGHY140035
Propargyl-PEG24-amine is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Click chemistry reagent
  • Contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups