Learn More
Medchemexpress LLC Propargyl-PEG3-acid | 1347760-82-0 | 99.1% | 216.23 | C10H16O5 | 250 MG
SDP
Catalog No. 5000204188
Encompass_Preferred
Propargyl-PEG3-acid is a PEG-based bifunctional linker featuring a terminal propargyl (alkyne) group and a carboxylic acid; it is used as a non-cleavable three-unit PEG linker for antibody-drug conjugates and as a PEG linker for PROTACs, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAC) click chemistry.
  • Non-cleavable three-unit PEG linker suitable for ADC and PROTAC synthesis.
  • Terminal alkyne enables copper-catalyzed azide-alkyne cycloaddition for conjugation to azide-containing molecules.
  • High purity appropriate for research-scale bioconjugation workflows.
  • Available in multiple pack sizes to support small-scale and larger preparative work.
Catalog No. 50-002-04188 Supplier Medchemexpress LLC Supplier No. HY126975250MG
Propargyl-PEG3-acid is a PEG-based bifunctional linker featuring a terminal propargyl (alkyne) group and a carboxylic acid; it is used as a non-cleavable three-unit PEG linker for antibody-drug conjugates and as a PEG linker for PROTACs, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAC) click chemistry.
  • Non-cleavable three-unit PEG linker suitable for ADC and PROTAC synthesis.
  • Terminal alkyne enables copper-catalyzed azide-alkyne cycloaddition for conjugation to azide-containing molecules.
  • High purity appropriate for research-scale bioconjugation workflows.
  • Available in multiple pack sizes to support small-scale and larger preparative work.