Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Propargyl-Peg3-Acid

Catalog No. 90156550MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-156-550MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-156-550MG Supplier Sigma Aldrich Supplier No. 90156550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

This is a crosslinker with an alkyne group on one end and a carboxyl group on the other end. The alkyne can be used to attach to an azide using copper-catalyzed click chemistry. The compound contains three PEG units to help improve solubility.PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Specifications

CAS 51641-96-4
Molecular Formula C24H31N3O10
Linear Formula C24H31N3O10
Quantity 50 mg
Synonym 1-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)-3,6,9,12-tetraoxapentadecan-15-oic acid; Crosslinker-E3 Ligase ligand conjugate; Protein degrader building block for PROTAC(R) research; Template for synthesis of targeted protein degrader
Molecular Weight (g/mol) 521.52 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.