Learn More
Medchemexpress LLC Propargyl-PEG3-amine | 932741-19-0 | 98.0% | 187.24 | 100 MG
SDP
Catalog No. 5000347975
Encompass_Preferred
Propargyl-PEG3-amine is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is also a click chemistry reagent that contains an alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups
Catalog No. 50-003-47975 Supplier Medchemexpress LLC Supplier No. HY140033100MG
Propargyl-PEG3-amine is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is also a click chemistry reagent that contains an alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups