Learn More
Medchemexpress LLC Propargyl-PEG3-amine | 932741-19-0 | ≥98.0% | 187.24 | 250 MG
SDP
Catalog No. 5000349397
Encompass_Preferred
Propargyl-PEG3-amine is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • PEG-based PROTAC linker
  • Functions as a click chemistry reagent
  • Contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Reacts with molecules containing azide groups
  • Used in the synthesis of PROTACs
Catalog No. 50-003-49397 Supplier Medchemexpress LLC Supplier No. 250MGHY140033
Propargyl-PEG3-amine is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • PEG-based PROTAC linker
  • Functions as a click chemistry reagent
  • Contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Reacts with molecules containing azide groups
  • Used in the synthesis of PROTACs