Learn More
Medchemexpress LLC Propargyl-peg3-bromide | 203740-63-0 | 97.66% | 251.12 | 100 MG
SDP
Catalog No. 5000349768
Encompass_Preferred
Propargyl-PEG3-bromide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is also a click chemistry reagent containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (Cuaac)
  • Reacts with molecules containing azide groups
Catalog No. 50-003-49768 Supplier Medchemexpress LLC Supplier No. HY140036100MG
Propargyl-PEG3-bromide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is also a click chemistry reagent containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (Cuaac)
  • Reacts with molecules containing azide groups