Learn More
Medchemexpress LLC Propargyl-PEG3-bromide | 203740-63-0 | 97.7% | 251.12 | 1 G
SDP
Catalog No. 5000347976
Encompass_Preferred
Propargyl-PEG3-bromide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It also functions as a click chemistry reagent, containing an alkyne group capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that possess azide groups. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Click chemistry
  • Alkynes
  • PROTAC synthesis
  • Disease areas: Cancer, Cancer targeted therapy
  • PROTAC linkers
Catalog No. 50-003-47976 Supplier Medchemexpress LLC Supplier No. HY1400361G
Propargyl-PEG3-bromide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It also functions as a click chemistry reagent, containing an alkyne group capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that possess azide groups. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Click chemistry
  • Alkynes
  • PROTAC synthesis
  • Disease areas: Cancer, Cancer targeted therapy
  • PROTAC linkers