Learn More
Medchemexpress LLC Propargyl-PEG3-bromide | 203740-63-0 | 97.7% | 251.12 | 500 MG
SDP
Catalog No. 5000351373
Encompass_Preferred
Propargyl-PEG3-bromide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with azide-containing molecules.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
Catalog No. 50-003-51373 Supplier Medchemexpress LLC Supplier No. HY140036500MG
Propargyl-PEG3-bromide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with azide-containing molecules.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups