Learn More
Medchemexpress LLC Propargyl-PEG3-CH2COOH | 1694731-93-5 | 100 MG
SDP
Catalog No. 5000339554
Encompass_Preferred
Propargyl-PEG3-CH2COOH is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Functions as a PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Acts as a click chemistry reagent
  • Contains an alkyne group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
Catalog No. 50-003-39554 Supplier Medchemexpress LLC Supplier No. HY130563100MG
Propargyl-PEG3-CH2COOH is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Functions as a PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Acts as a click chemistry reagent
  • Contains an alkyne group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups