Learn More
Medchemexpress LLC Propargyl-PEG3-CH2COOH | 1694731-93-5 | 98.0% | 25 MG
SDP
Catalog No. 5000341450
Encompass_Preferred
Propargyl-PEG3-CH2COOH is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein, exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Click chemistry reagent with an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-41450 Supplier Medchemexpress LLC Supplier No. 25MGHY130563
Propargyl-PEG3-CH2COOH is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein, exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Click chemistry reagent with an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system to selectively degrade target proteins