Learn More
Medchemexpress LLC Propargyl-PEG3-CH2COOH | 1694731-93-5 | 98.0% | 50 MG
SDP
Catalog No. 5000340205
Encompass_Preferred
Propargyl-PEG3-CH2COOH is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that contain Azide groups.
  • PEG-based PROTAC linker.
  • Functions as a click chemistry reagent.
  • Contains an Alkyne group.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
Catalog No. 50-003-40205 Supplier Medchemexpress LLC Supplier No. HY13056350MG
Propargyl-PEG3-CH2COOH is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that contain Azide groups.
  • PEG-based PROTAC linker.
  • Functions as a click chemistry reagent.
  • Contains an Alkyne group.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.