Learn More
Medchemexpress LLC Propargyl-PEG3-methane | 89635-82-5 | 98.0% | 202.25 | 25 G
SDP
Catalog No. 5000351871
Encompass_Preferred
Propargyl-PEG3-methane is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules possessing Azide groups. For research use only. Not sold to patients.
  • Click chemistry reagent
  • Alkynes
  • PROTAC synthesis
  • PROTAC linkers
  • Used in cancer research
  • Applied in cancer targeted therapy
Catalog No. 50-003-51871 Supplier Medchemexpress LLC Supplier No. 25GHY140059
Propargyl-PEG3-methane is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules possessing Azide groups. For research use only. Not sold to patients.
  • Click chemistry reagent
  • Alkynes
  • PROTAC synthesis
  • PROTAC linkers
  • Used in cancer research
  • Applied in cancer targeted therapy