Learn More
Medchemexpress LLC Propargyl-PEG3-methane | 89635-82-5 | MFCD27977518 | 98.0% | 202.25 | 500 MG
SDP
Catalog No. 5000347979
Encompass_Preferred
Propargyl-PEG3-methane is a PEG-based PROTAC linker, designed for use in the synthesis of Proteolysis Targeting Chimeras (PROTACs). It functions as a versatile click chemistry reagent, facilitating various bioconjugation and labeling applications.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Facilitates bioconjugation and labeling
Catalog No. 50-003-47979 Supplier Medchemexpress LLC Supplier No. HY140059500MG
Propargyl-PEG3-methane is a PEG-based PROTAC linker, designed for use in the synthesis of Proteolysis Targeting Chimeras (PROTACs). It functions as a versatile click chemistry reagent, facilitating various bioconjugation and labeling applications.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Facilitates bioconjugation and labeling