Learn More
Medchemexpress LLC Propargyl-PEG3-OCH2-Boc | 888010-02-4 | 302.36 | 100 MG
SDP
Catalog No. 5000349853
Encompass_Preferred
Propargyl-PEG3-OCH2-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
  • Relevant for cancer and cancer targeted therapy research
  • PROTAC linker classification
Catalog No. 50-003-49853 Supplier Medchemexpress LLC Supplier No. HY140031100MG
Propargyl-PEG3-OCH2-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
  • Relevant for cancer and cancer targeted therapy research
  • PROTAC linker classification