Learn More
Medchemexpress LLC Propargyl-PEG3-OCH2-Boc | 888010-02-4 | 302.36 | 25 MG
SDP
Catalog No. 5000349988
Encompass_Preferred
Propargyl-PEG3-OCH2-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Used in the synthesis of PROTACs
  • For research use only
Catalog No. 50-003-49988 Supplier Medchemexpress LLC Supplier No. 25MGHY140031
Propargyl-PEG3-OCH2-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Used in the synthesis of PROTACs
  • For research use only