Learn More
Medchemexpress LLC Propargyl-PEG3-OCH2-Boc | 888010-02-4 | 302.36 | 250 MG
SDP
Catalog No. 5000349396
Encompass_Preferred
Propargyl-PEG3-OCH2-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is also a click chemistry reagent containing an Alkyne group, capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent containing an alkyne group
  • Capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups
  • Utilizes two different ligands connected by a linker; one ligand for an E3 ubiquitin ligase and another for the target protein
  • Employs the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-49396 Supplier Medchemexpress LLC Supplier No. 250MGHY140031
Propargyl-PEG3-OCH2-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is also a click chemistry reagent containing an Alkyne group, capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent containing an alkyne group
  • Capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups
  • Utilizes two different ligands connected by a linker; one ligand for an E3 ubiquitin ligase and another for the target protein
  • Employs the intracellular ubiquitin-proteasome system to selectively degrade target proteins