Learn More
Medchemexpress LLC Propargyl-PEG3-OCH2-Boc | 888010-02-4 | 302.36 | 50 MG
SDP
Catalog No. 5000351372
Encompass_Preferred
Propargyl-PEG3-OCH2-Boc is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Alkyne group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. This compound is strictly for research use and is not intended for patient treatment.
  • PEG-based PROTAC linker for PROTAC synthesis
  • Click chemistry reagent with Alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Reacts with molecules containing Azide groups
  • Chemical formula: C15H26O6
  • Intended for research use only
Catalog No. 50-003-51372 Supplier Medchemexpress LLC Supplier No. HY14003150MG
Propargyl-PEG3-OCH2-Boc is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Alkyne group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. This compound is strictly for research use and is not intended for patient treatment.
  • PEG-based PROTAC linker for PROTAC synthesis
  • Click chemistry reagent with Alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Reacts with molecules containing Azide groups
  • Chemical formula: C15H26O6
  • Intended for research use only