Learn More
Medchemexpress LLC Propargyl-PEG4-5-nitrophenyl carbonate | 1422540-81-5 | 98.19% | 397.38 | 25 MG
SDP
Catalog No. 5000349412
Encompass_Preferred
Propargyl-PEG4-5-nitrophenyl carbonate is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It acts as a click chemistry reagent, possessing an Alkyne group that enables it to engage in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. PROTACs consist of two distinct ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein, facilitating the selective degradation of target proteins via the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
Catalog No. 50-003-49412 Supplier Medchemexpress LLC Supplier No. 25MGHY140048
Propargyl-PEG4-5-nitrophenyl carbonate is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It acts as a click chemistry reagent, possessing an Alkyne group that enables it to engage in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. PROTACs consist of two distinct ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein, facilitating the selective degradation of target proteins via the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups