Learn More
Medchemexpress LLC Propargyl-PEG4-5-nitrophenyl carbonate | 1422540-81-5 | 98.2% | 397.38 | 50 MG
SDP
Catalog No. 5000347977
Encompass_Preferred
Propargyl-PEG4-5-nitrophenyl carbonate is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It also functions as a click chemistry reagent, featuring an Alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Click chemistry reagent
  • Features an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
  • Exploits ubiquitin-proteasome system for selective protein degradation
Catalog No. 50-003-47977 Supplier Medchemexpress LLC Supplier No. HY14004850MG
Propargyl-PEG4-5-nitrophenyl carbonate is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It also functions as a click chemistry reagent, featuring an Alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Click chemistry reagent
  • Features an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
  • Exploits ubiquitin-proteasome system for selective protein degradation