Learn More
Medchemexpress LLC Propargyl-PEG4-acid | 1415800-32-6 | 97.0% | 1 G
SDP
Catalog No. 5000338734
Encompass_Preferred
Propargyl-PEG4-acid is a PEG-based PROTAC linker primarily used in the synthesis of BTK-IAP PROTACs. It also functions as a click chemistry reagent, containing an Alkyne group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups. These PROTACs act as stoichiometric degraders, leading to the degradation of BTK protein by recruiting IAP E3 ligases.
  • PEG-based PROTAC linker
  • Used in the synthesis of BTK-IAP PROTACs
  • Functions as a click chemistry reagent
  • Contains an Alkyne group for CuAAc reactions
  • Enables degradation of BTK protein
Catalog No. 50-003-38734 Supplier Medchemexpress LLC Supplier No. HY1304811G
Propargyl-PEG4-acid is a PEG-based PROTAC linker primarily used in the synthesis of BTK-IAP PROTACs. It also functions as a click chemistry reagent, containing an Alkyne group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups. These PROTACs act as stoichiometric degraders, leading to the degradation of BTK protein by recruiting IAP E3 ligases.
  • PEG-based PROTAC linker
  • Used in the synthesis of BTK-IAP PROTACs
  • Functions as a click chemistry reagent
  • Contains an Alkyne group for CuAAc reactions
  • Enables degradation of BTK protein