Learn More
Medchemexpress LLC Propargyl-PEG4-acid | 1415800-32-6 | 97.0% | 25 G
SDP
Catalog No. 5000341475
Encompass_Preferred
Propargyl-PEG4-acid is a PEG-based PROTAC linker used in the synthesis of BTK-IAP PROTACs, including Ibrutinib-based PROTAC 2 and its analogue PROTAC 3. PROTAC 3 has demonstrated BTK degradation with a DC50 of 200 nM in THP-1 cells. This compound also serves as a click chemistry reagent, featuring an Alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules. It is intended for research use only.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of BTK-IAP PROTACs
  • Functions as a click chemistry reagent
  • Possesses an Alkyne group for copper-catalyzed azide-alkyne cycloaddition with azide-containing molecules
  • Demonstrates BTK degradation in THP-1 cells
  • Intended for research use only
Catalog No. 50-003-41475 Supplier Medchemexpress LLC Supplier No. 25GHY130481
Propargyl-PEG4-acid is a PEG-based PROTAC linker used in the synthesis of BTK-IAP PROTACs, including Ibrutinib-based PROTAC 2 and its analogue PROTAC 3. PROTAC 3 has demonstrated BTK degradation with a DC50 of 200 nM in THP-1 cells. This compound also serves as a click chemistry reagent, featuring an Alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules. It is intended for research use only.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of BTK-IAP PROTACs
  • Functions as a click chemistry reagent
  • Possesses an Alkyne group for copper-catalyzed azide-alkyne cycloaddition with azide-containing molecules
  • Demonstrates BTK degradation in THP-1 cells
  • Intended for research use only