Learn More
Medchemexpress LLC Propargyl-PEG4-acid | 1415800-32-6 | 97.0% | 500 MG
SDP
Catalog No. 5000340833
Encompass_Preferred
Propargyl-PEG4-acid is a PEG-based PROTAC linker that can be used in the synthesis of BTK-IAP PROTACs Ibrutinib-based PROTAC 2 and an analogue PROTAC 3. PROTAC 3 causes BTK degradation with a DC50 of 200 nM in THP-1 cells.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of BTK-IAP PROTACs
  • Click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups
Catalog No. 50-003-40833 Supplier Medchemexpress LLC Supplier No. HY130481500MG
Propargyl-PEG4-acid is a PEG-based PROTAC linker that can be used in the synthesis of BTK-IAP PROTACs Ibrutinib-based PROTAC 2 and an analogue PROTAC 3. PROTAC 3 causes BTK degradation with a DC50 of 200 nM in THP-1 cells.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of BTK-IAP PROTACs
  • Click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups