Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-PEG4-amine 1g | 1013921-36-2 | 1 G
SDP

Catalog No. 5000228211
Encompass_Preferred

Propargyl-PEG4-amine is an alkyne-functional tetraethylene glycol amine used as a linker for bioconjugation and PROTAC synthesis. It provides an alkyne handle for copper-catalyzed azide-alkyne cycloaddition and a terminal primary amine for further coupling reactions.

  • Provides an alkyne handle for copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • Contains a tetraethylene glycol spacer to improve solubility and flexibility.
  • Offers a terminal primary amine for amide bond formation and other coupling chemistries.
  • High purity suitable for synthesis and conjugation (99.7%).
  • Molecular weight 231.29 g/mol; formula C11H21NO4.

Catalog No. 50-002-28211 Supplier Medchemexpress LLC Supplier No. HY1146701G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-PEG4-amine is an alkyne-functional tetraethylene glycol amine used as a linker for bioconjugation and PROTAC synthesis. It provides an alkyne handle for copper-catalyzed azide-alkyne cycloaddition and a terminal primary amine for further coupling reactions.

  • Provides an alkyne handle for copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • Contains a tetraethylene glycol spacer to improve solubility and flexibility.
  • Offers a terminal primary amine for amide bond formation and other coupling chemistries.
  • High purity suitable for synthesis and conjugation (99.7%).
  • Molecular weight 231.29 g/mol; formula C11H21NO4.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.