Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-PEG4-amine 5g | 1013921-36-2 | 5 G
SDP

Catalog No. 5000229256
Encompass_Preferred

Propargyl-PEG4-amine is an alkyne-terminated PEG4 linker with a primary amine, used in PROTAC linker assembly, click chemistry, and bioconjugation workflows. It enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) and amide bond formation for attaching payloads or linkers.

  • Contains terminal alkyne for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Primary amine enables amide coupling to activated carboxylic acids or NHS esters.
  • Peg4 spacer provides flexibility and improved solubility in aqueous systems.
  • Molecular weight 231.29 g/mol and formula C11H21NO4.
  • High purity suitable for chemical biology and linker synthesis applications.

Catalog No. 50-002-29256 Supplier Medchemexpress LLC Supplier No. HY1146705G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-PEG4-amine is an alkyne-terminated PEG4 linker with a primary amine, used in PROTAC linker assembly, click chemistry, and bioconjugation workflows. It enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) and amide bond formation for attaching payloads or linkers.

  • Contains terminal alkyne for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Primary amine enables amide coupling to activated carboxylic acids or NHS esters.
  • Peg4 spacer provides flexibility and improved solubility in aqueous systems.
  • Molecular weight 231.29 g/mol and formula C11H21NO4.
  • High purity suitable for chemical biology and linker synthesis applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.