Learn More
Medchemexpress LLC Propargyl-PEG4-Boc | 1355197-66-8 | 316.39 | 1 G
SDP
Catalog No. 5000349166
Encompass_Preferred
Propargyl-PEG4-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is a click chemistry reagent, containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an Alkyne group
  • Can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups
Catalog No. 50-003-49166 Supplier Medchemexpress LLC Supplier No. HY1400281G
Propargyl-PEG4-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is a click chemistry reagent, containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an Alkyne group
  • Can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups