Learn More
Medchemexpress LLC Propargyl-PEG4-Boc | 1355197-66-8 | 316.39 | 100 MG
SDP
Catalog No. 5000349962
Encompass_Preferred
Propargyl-PEG4-Boc is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group, which allows it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that contain Azide groups.
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an alkyne group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules
Catalog No. 50-003-49962 Supplier Medchemexpress LLC Supplier No. HY140028100MG
Add to Cart
Add to Cart
Propargyl-PEG4-Boc is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group, which allows it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that contain Azide groups.
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an alkyne group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules