Learn More
Medchemexpress LLC Propargyl-PEG4-Boc | 1355197-66-8 | 316.39 | 250 MG
SDP
Catalog No. 5000348256
Encompass_Preferred
Propargyl-PEG4-Boc is a PEG-based PROTAC linker designed for the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Alkyne group that readily undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. This reagent facilitates the creation of PROTACs, which are compounds that exploit the ubiquitin-proteasome system to degrade target proteins.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains an Alkyne group for CuAAc reactions
  • Used in the synthesis of PROTACs
Catalog No. 50-003-48256 Supplier Medchemexpress LLC Supplier No. 250MGHY140028
Propargyl-PEG4-Boc is a PEG-based PROTAC linker designed for the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Alkyne group that readily undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. This reagent facilitates the creation of PROTACs, which are compounds that exploit the ubiquitin-proteasome system to degrade target proteins.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains an Alkyne group for CuAAc reactions
  • Used in the synthesis of PROTACs