Learn More
Medchemexpress LLC Propargyl-PEG4-Br | 1308299-09-3 | 1 G
SDP
Catalog No. 5000340218
Encompass_Preferred
Propargyl-PEG4-Br is a PEG-based PROTAC linker and a non-cleavable ADC linker. It is used in the synthesis of PROTACs and antibody-drug conjugates (ADCs). This click chemistry reagent contains an Alkyne group, allowing it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAC) with Azide group-containing molecules. PROTACs selectively degrade target proteins using the ubiquitin-proteasome system, while ADCs attach cytotoxins to antibodies via linkers.
  • Functions as a PEG-based PROTAC linker
  • Enables synthesis of PROTACs
  • Acts as a non-cleavable ADC linker
  • Facilitates synthesis of antibody-drug conjugates (ADCs)
  • Serves as a click chemistry reagent
  • Contains an alkyne group for reactions
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAC)
  • Reacts with azide group-containing molecules
  • Features PEG5 structure
Catalog No. 50-003-40218 Supplier Medchemexpress LLC Supplier No. HY1305911G
Add to Cart
Add to Cart
Propargyl-PEG4-Br is a PEG-based PROTAC linker and a non-cleavable ADC linker. It is used in the synthesis of PROTACs and antibody-drug conjugates (ADCs). This click chemistry reagent contains an Alkyne group, allowing it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAC) with Azide group-containing molecules. PROTACs selectively degrade target proteins using the ubiquitin-proteasome system, while ADCs attach cytotoxins to antibodies via linkers.
  • Functions as a PEG-based PROTAC linker
  • Enables synthesis of PROTACs
  • Acts as a non-cleavable ADC linker
  • Facilitates synthesis of antibody-drug conjugates (ADCs)
  • Serves as a click chemistry reagent
  • Contains an alkyne group for reactions
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAC)
  • Reacts with azide group-containing molecules
  • Features PEG5 structure