Learn More
Medchemexpress LLC Propargyl-PEG4-Br | 1308299-09-3 | 5 G
SDP
Catalog No. 5000341052
Encompass_Preferred
Propargyl-PEG4-Br is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a non-cleavable ADC linker utilized in the synthesis of antibody-drug conjugates (ADCs).
  • Functions as a PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Acts as a non-cleavable ADC linker
  • Utilized in the synthesis of antibody-drug conjugates (ADCs)
  • Serves as a click chemistry reagent
  • Contains an Alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups
Catalog No. 50-003-41052 Supplier Medchemexpress LLC Supplier No. HY1305915G
Propargyl-PEG4-Br is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a non-cleavable ADC linker utilized in the synthesis of antibody-drug conjugates (ADCs).
  • Functions as a PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Acts as a non-cleavable ADC linker
  • Utilized in the synthesis of antibody-drug conjugates (ADCs)
  • Serves as a click chemistry reagent
  • Contains an Alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups