Learn More
Medchemexpress LLC Propargyl-PEG4-Br | 1308299-09-3 | 97.0% | 250 MG
SDP
Catalog No. 5000341509
Encompass_Preferred
Propargyl-PEG4-Br is a PEG-based PROTAC linker used in the synthesis of PROTACs. It also functions as a non-cleavable ADC linker for antibody-drug conjugates (ADCs). This compound is a click chemistry reagent, featuring an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAC) with azide-containing molecules.
  • PEG-based PROTAC linker
  • Non-cleavable ADC linker
  • Click chemistry reagent
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition
Catalog No. 50-003-41509 Supplier Medchemexpress LLC Supplier No. 250MGHY130591
Propargyl-PEG4-Br is a PEG-based PROTAC linker used in the synthesis of PROTACs. It also functions as a non-cleavable ADC linker for antibody-drug conjugates (ADCs). This compound is a click chemistry reagent, featuring an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAC) with azide-containing molecules.
  • PEG-based PROTAC linker
  • Non-cleavable ADC linker
  • Click chemistry reagent
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition