Learn More
Medchemexpress LLC Propargyl-PEG4-CH2-methyl ester | 1872433-63-0 | 250 MG
SDP
Catalog No. 5000342625
Encompass_Preferred
Propargyl-PEG4-CH2-methyl ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an Alkyne group, which can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Peg-based protac linker
  • Click chemistry reagent
  • Contains an alkyne group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups
Catalog No. 50-003-42625 Supplier Medchemexpress LLC Supplier No. 250MGHY130293
Propargyl-PEG4-CH2-methyl ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an Alkyne group, which can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Peg-based protac linker
  • Click chemistry reagent
  • Contains an alkyne group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups